Would you like to log in again?
Pierce™ 96 Well Spin Filter Plates, Thermo Scientific
Supplier: Thermo Scientific
Pierce
16111
Thermo Scientific Pierce™ Glutathione spin plates are convenient, ready to use pre-dispensed filter plates for the efficient purification of GST-tagged proteins from bacterial, mammalian and baculovirus-infected cells
  • Glutathione is immobilized through its central sulfhydryl onto 6% crosslinked agarose resin
  • Binds at least 40 mg of purified recombinant GST protein per mL of resin
  • Consistently purifies at least 10 mg of GST-tagged protein per mL of resin with greater than 90% purity
  • Resin can be reused at least five times without reduction in performance
The Pierce™ Glutathione spin plates are compatible with centrifugation and vacuum manifold systems for manual or automated purification. The plates enable fast, consistent well-to-well and plate-to-plate reproducibility for small-scale high throughput separations.
Delivery information: Includes two filter plates with each well containing 50 µL of resin bed (100 µL of 50% resin slurry) and 6X wash/collection plates.
ORDER
Expand All / Collapse All
Description Size Supplier No. VWR Catalog Number Unit Price Quantity
Glutathione spin plates 2 plates 16111 PI16111 Each Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
ACCESSORIES
Expand All / Collapse All
Collection Plates and Balance Plates for Pierce™ 96-Well Filter Spin Plates
Size Supplier No. VWR Catalog Number Unit Price Quantity
1 plate 45205 PI45205 Each Retrieving Restricted
6 plates 89813 PI89813 Each Retrieving Restricted
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty