SP Wilmad-LabGlass 90° Bend Drying Tube, SP Industries
Supplier: SP Industries
ML-4095-600
82003-074EA 43.47 USD
82003-074
SP Wilmad-LabGlass 90° Bend Drying Tube, SP Industries
This borosilicate glass drying tube is used to protect moisture sensitive reactions from water vapor in the atmosphere, while permitting the reaction system to remain open to the atmosphere.
With 90° bend at end closest to inner Standard Taper 14/10 joint.