Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DualFUSION Vials, Wheaton

Supplier: DWK Life Sciences
W266110 W266104 W266108 W266106
DualFUSION vials provide customers with the best of both worlds: lightweight, shatter-resistant properties of plastic and an inert inner lining with glass barrier properties.

  • No mechanism of delamination
  • Low level of oxygen permeation
  • Anti counterfeiting technologies
  • Meets USP 788 and 85

DualFUSION combines the best properties of plastic and glass into one vial

DualFUSION vials are manufactured in a Class 7 cleanroom, resulting in a “ready-to-sterilize” vial. Vials are equipped with a 2D bar code, offering complete manufacturing traceability.

The new “FUSION” material lining offers protection against leachates, delamination, and gas permeation. The Cyclic Olefin Polymer (COP) shell provides users with the comfort of a break-resistant vial.



 

ORDER

Expand All / Collapse All
Capacity Diameter Height Cap Size Cap Type Supplier No. VWR Catalog Number Unit Price Quantity
5 mL 22 mm 40 mm 20 mm Crimp Finish W266104 75876-408 Case of 48 (5ml) Retrieving
10 mL 24 mm 45 mm 20 mm Crimp Finish W266108 75876-412 Case of 48 (10ml) Retrieving
5 mL 22 mm 40 mm 20 mm Crimp Finish W266106 75876-410 Case of 96 (5ml) Retrieving
10 mL 24 mm 45 mm 20 mm Crimp Finish W266110 75876-414 Case of 96 (10ml) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty