1-Octanol ≥99%, CHROMASOLV™ for HPLC, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Warning

Synonyms: n-Octanol

293245-1L
BJ293245-1LEA 230.71 USD
BJ293245-1L
1-Octanol ≥99%, CHROMASOLV™ for HPLC, Riedel-de Haën™
1-Octanol

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency

Formula: H₃C(CH₂)₆CH₂OH
MW: 130.23 g/mol
Boiling Pt: 194 °C (1.013 hPa)
Melting Pt: -15 °C
Density: 0.827 g/cm³ (25 °C)
Flash Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00002988
CAS Number: 111-87-5
EINECS: 203-917-6
https://us.vwr.com/stibo/web/std.lang.all/14/03/731403.jpg

Order Now

Specification Test Results

Identity (IR) complying
Assay (GC) Min. 99.0 %
Non-volatile matter Max. 0.004 %
Water (Karl Fischer) Max. 0.01 %
Absorbance at 400 nm Max. 0.01
Absorbance at 300 nm Max. 0.01
Absorbance at 250 nm Max. 0.05
Absorbance at 235 nm Max. 0.20
Absorbance at 225 nm Max. 0.50
Absorbance at 215 nm Max. 1.0
APHA Max. 10
Refractive Index n20/D 1.429 (lit.)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR