Tashiro’s indicator solution in ethanol, Fluka™
|
Danger
|
36083-6X1L
36083-6X250ML
BJ36083-1LCS
610.76
USD
BJ36083-1L
BJ36083-250ML
Tashiro’s indicator solution in ethanol, Fluka™
Tashiro’s indicator solution
Boiling Pt:
78 °C (1.013 hPa) Density: 0.79 g/cm³ (20 °C) Flash Pt: 12 °C Storage Temperature: Ambient |
MDL Number:
MFCD00148563 EINECS: 200-578-6 UN: 1170 ADR: 3,II |
Specification Test Results
Change range (pH 5-6) | complying |