Safety-Coated Glass Jugs, Amber, Environmetal Express
Supplier: Environmental Express
APC2800
89202-861CS 225.07 USD
89202-861
Safety-Coated Glass Jugs, Amber, Environmetal Express
Bottles Glass Bottles
Glass jug is PVC safety-coated and supplied with a 38 mm black closure with PTFE liner.