    Sort by:
  • Search Within Results
Soap solution according to pellet
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Catalog Number: (BJ31089-5L)
Supplier:  Honeywell Research Chemicals
Description:   1 ml = 0.001 g CaO in 100 ml water = 1 German degree of hardness