Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

High Performance WebSeal™ Plates and Glass Coated Microplates, Thermo Scientific

Supplier: Thermo Scientific
60180-P300 60180-P310 60180-P302 60180-P313 60180-P312 60180-P304 60180-P303 60180-P306 60180-P308
High quality polypropylene microplates are coated with a 200 nm thick layer of silicone dioxide

Chemical resistance is similar to glass, while retaining the advantages of polypropylene construction.




 

ORDER

Expand All / Collapse All
Description Volume Height Well Shape Supplier No. VWR Catalog Number Unit Price Quantity
96-Well Microplate, Round Well Total: 300 µL, Working: 250 µL 14.6 mm U-Shape, 7.2 mm Diameter 60180-P300 10036-256 Pack of 10 Retrieving
96-Well Microplate, Round Well Total: 220 µL, Working: 190 µL 14.6 mm V-Shape, 7.2 mm Diameter 60180-P302 10036-260 Pack of 10 Retrieving
96-Well Microplate, Round Well Total: 370 µL, Working: 300 µL 14.6 mm Flat Bottom, 7.2 mm Diameter 60180-P304 10036-264 Pack of 10 Retrieving
96-Well Microplate, Round Well Total: 220 µL, Working: 190 µL 14.6 mm V-Shape, 7.2 mm Diameter 60180-P303 10036-262 Case of 100 Retrieving
96-Well Mid Height Plate, Round Well Total: 1.2 mL, Working: 1.0 mL 41.5 mm U-Shape, 7.2 mm Diameter 60180-P306 10036-268 Pack of 10 Retrieving
96-Well Deep Well Plate, Square Well Total: 2.4 mL, Working: 2.0 mL 44 mm V-Shape, 8 mm Diameter 60180-P308 10036-272 Pack of 10 Retrieving
384-Well Microplate Total: 120 µL, Working: 90 µL 14.4 mm Square-Rounded, Flat, Microtiter Plate 60180-P310 10036-276 Pack of 10 Retrieving
384-Well Microplate Total: 240 µL 22 mm Square-Rounded, U-Shape, Deep Well Plate 60180-P312 10036-280 Pack of 6 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Material PP, glass coated

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty