Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chemical OSHA Danger Signs, Arsenic, NMC

Supplier: NATIONAL MARKER COMP.
D562RB D562PB D562AB
OSHA signs are the perfect way to alert your employees and guests when a hazard is present in your facility.

  • Ideal for indoor and outdoor labeling
  • Available in Alum, PS vinyl, Fiber glass and Rigid plastic (thick polystyrene) materials
  • .040 alum is weatherproof, non-corrosive and has baked-on enamel on both sides
  • .004 PS vinyl material has pressure sensitive adhesive backing
  • .095 fiber glass material is rigid, shatter-proof and dent resistant
  • .050 rigid plastic material has radius corners and pre-drilled corner mounting holes

Keep your facility safe, buy an OSHA regulated sign today and prevent an accident tomorrow. Each sign header reads "Danger".

Certifications: Comply with OSHA 29 CFR 1910.



 

ORDER

Expand All / Collapse All
Size Material Legend Supplier No. VWR Catalog Number Unit Price Quantity
25.4 x 35.56 cm (10 x 14") .040 Alum Inorganic Arsenic Cancer Hazard Authorized Personnel Only No Smoking Or Eating Respirator Required D562AB 94010-870 Each Retrieving
25.4 x 35.56 cm (10 x 14") PS Vinyl Inorganic Arsenic Cancer Hazard Authorized Personnel Only No Smoking Or Eating Respirator Required D562PB 94010-872 Each Retrieving
25.4 x 35.56 cm (10 x 14") Rigid Plastic Inorganic Arsenic Cancer Hazard Authorized Personnel Only No Smoking Or Eating Respirator Required D562RB 94010-874 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty