Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chemical OSHA Danger Signs, Poison, NMC

Supplier: NATIONAL MARKER COMP.
D561AB D463PB D440PB D463RB D440RB D319PB D82AB D602AB D601AB D440R D463AB D561RB D440P D561PB D440AB D463EB D440EB D601RB D601PB D602RB D282P D282PB D602PB
OSHA signs are the perfect way to alert your employees and guests when a hazard is present in your facility.

  • Ideal for indoor and outdoor labeling
  • Available in Alum, PS vinyl, Fiber glass and Rigid plastic (thick polystyrene) materials
  • .040 alum is weatherproof, non-corrosive and has baked-on enamel on both sides
  • .004 PS vinyl material has pressure sensitive adhesive backing
  • .095 fiber glass material is rigid, shatter-proof and dent resistant
  • .050 rigid plastic material has radius corners and pre-drilled corner mounting holes

Keep your facility safe, buy an OSHA regulated sign today and prevent an accident tomorrow. Each sign header reads "Danger".

Certifications: Comply with OSHA 29 CFR 1910.



 

ORDER

Expand All / Collapse All
Size Material Legend Supplier No. VWR Catalog Number Unit Price Quantity
25.4 x 35.56 cm (10 x 14") .040 Alum Poison D463AB 94010-148 Each Retrieving
25.4 x 35.56 cm (10 x 14") Fiberglass Poison D463EB 94010-150 Each Retrieving
25.4 x 35.56 cm (10 x 14") PS Vinyl Poison D463PB 94010-152 Each Retrieving
25.4 x 35.56 cm (10 x 14") Rigid Plastic Poison D463RB 94010-154 Each Retrieving
25.4 x 35.56 cm (10 x 14") .040 Alum Inhalation Hazard Do Not Breathe Vapors D561AB 94010-864 Each Retrieving
25.4 x 35.56 cm (10 x 14") PS Vinyl Inhalation Hazard Do Not Breathe Vapors D561PB 94010-866 Each Retrieving
25.4 x 35.56 cm (10 x 14") Rigid Plastic Inhalation Hazard Do Not Breathe Vapors D561RB 94010-868 Each Retrieving
17.78 x 25.4 cm (7 x 10") PS Vinyl H2S May Be Present D282P 94008-974 Each Retrieving
25.4 x 35.56 cm (10 x 14") PS Vinyl H2S May Be Present D282PB 94008-976 Each Retrieving
25.4 x 35.56 cm (10 x 14") PS Vinyl Toxic Chemicals D319PB 94009-144 Each Retrieving
25.4 x 35.56 cm (10 x 14") .040 Alum Poison D601AB 94011-150 Each Retrieving
25.4 x 35.56 cm (10 x 14") PS Vinyl Poison D601PB 94011-152 Each Retrieving
25.4 x 35.56 cm (10 x 14") Rigid Plastic Poison D601RB 94011-154 Each Retrieving
25.4 x 35.56 cm (10 x 14") .040 Alum Poisonous Gas Hydrogen Sulfide D602AB 94011-156 Each Retrieving
25.4 x 35.56 cm (10 x 14") PS Vinyl Poisonous Gas Hydrogen Sulfide D602PB 94011-158 Each Retrieving
25.4 x 35.56 cm (10 x 14") Rigid Plastic Poisonous Gas Hydrogen Sulfide D602RB 94011-160 Each Retrieving
25.4 x 35.56 cm (10 x 14") .040 Alum Poison Gas D82AB 94011-436 Each Retrieving
25.4 x 35.56 cm (10 x 14") Alum H2S Poisonous Gas D440AB 94009-932 Each Retrieving
25.4 x 35.56 cm (10 x 14") Fiberglass H2S Poisonous Gas D440EB 94009-934 Each Retrieving
17.78 x 25.4 cm (7 x 10") PS Vinyl H2S Poisonous Gas D440P 94009-936 Each Retrieving
25.4 x 35.56 cm (10 x 14") PS Vinyl H2S Poisonous Gas D440PB 94009-938 Each Retrieving
17.78 x 25.4 cm (7 x 10") Rigid Plastic H2S Poisonous Gas D440R 94009-940 Each Retrieving
25.4 x 35.56 cm (10 x 14") Rigid Plastic H2S Poisonous Gas D440RB 94009-942 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping