Urea
Supplier: Promega Corporation
Synonyms:
Carbonyl diamide
, Carbamide
, Carbonyldiamide
, Diaminomethanone
V3175
V3171
PAV3175EA
257.27
USD
PAV3175
PAV3171
Urea
Urea
Urea is a protein denaturant. Urea is qualified for use as the denaturing component in polyacrylamide gels.
Form: Fine, white, free-flowing pastilles
Can be stored at 15°C to 30°C (Protect from moisture)
Formula:
NH₂CONH₂ MW: 60.06 g/mol |
MDL Number:
MFCD00008022 CAS Number: 57-13-6 |
Specification Test Results
Purity | ≥99.0% |
Melting Point | 132–135°C |
A280 at 8M in water | ≤0.10 |
Chloride | ≤0.0005% |
Heavy Metals | ≤0.001% |
Iron | ≤0.001% |
Cyanate | none detected |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...