    Sort by:
  • Search Within Results
Fmoc-(Dmb)Leu-OH
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Supplier:  Bachem Americas
Description:   Derivative for the Fmoc-SPPS of backbone-protected peptides. Introduction of Dmb at appropriate positions prevents aggregation of the growing peptide ...