    Sort by:
  • Search Within Results
Fmoc-δ-azido-Nva-OH
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Bachem Americas
Description:   Ornithine-derived building block for "click chemistry" and other reactions involving azides.The azido moiety may also be selectively converted into a...