Actin, Smooth Muscle IHC Positive Control Slides, Newcomer Supply

Supplier: Newcomer Supply
3003A 3003B
75876-828BX 72.31 USD
75876-828 75876-830
Actin, Smooth Muscle IHC Positive Control Slides, Newcomer Supply
Slides Microscopy Control Slides
Actin, smooth muscle, control slides contain a section of positive staining uterus.

  • Tissue Type: (+) Uterus
  • All control tissue is sectioned on Superfrost® Plus slides
  • Staining validation technical memo included in each set
  • All tissues are formalin fixed, paraffin embedded FFPE
  • All slides are air dried
  • IHC slides are cut at 4 µm
  • Standard tissue placement is mid-high on slide

Newcomer supply actin, smooth muscle, control slides are for the positive immunohistochemical staining of smooth muscle actin

Staining Results:
Actin, smooth muscle positive expression will stain brown cytoplasmic staining.
Nuclei will stain blue.

These positive control slides are intended to be used to verify histological techniques and reagent reactivity. These slides are to be used for the qualitative purpose of determining positive or negative results, and are not intended to be used for any quantitative purpose.

Myofibroblasts and myoepithelial cells will also positively react.

Ordering information: All controls are cut in serial sections.
10 set = 10 unstained slides and 1 stained slide.
98 set = 98 unstained slides and 2 stained slides. (First & Last Slides are Stained)
Reactivity of IHC control slides are guaranteed for one year from the date of receipt and should be revalidated after one year to verify continued reactivity.
Please note the following: Allow 2-3 weeks for delivery for large sets. In human tissue unless otherwise indicated. All tissues are formalin fixed, paraffin embedded FFPE. All slides are heat dried. Immunopathology slides are cut at 4 microns. Standard tissue placement is mid-range of slide.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR