Calcium Histo Positive Control Slides, Newcomer Supply

Supplier: Newcomer Supply
4100B 4100A
75876-974BX 426.08 USD
75876-974 75876-972
Calcium Histo Positive Control Slides, Newcomer Supply
Slides Microscopy Control Slides
Calcium control slides contain a section of positive staining placenta.

  • Tissue Type: (+) Placenta
  • All control tissue is sectioned on Superfrost® Plus slides
  • Staining validation technical memo included in each set
  • All tissues are formalin fixed, paraffin embedded FFPE
  • All slides are air dried
  • Histo slides are cut at 4 µm, unless otherwise specified
  • Standard tissue placement is mid-range of slide

Newcomer supply calcium control slides are for the positive histochemical staining of calcium or calcium salts in tissue sections.

Staining Results:
Calcium salts will stain black to brown/black.
Nuclei will stain red.
Cytoplasm will stain light pink.

These positive control slides are intended to be used to verify histological techniques and reagent reactivity. These slides are to be used for the qualitative purpose of determining positive or negative results, and are not intended to be used for any quantitative purpose.

Ordering information: All controls are cut in serial sections.
10 set = 10 unstained slides and 1 stained slide.
98 set = 98 unstained slides and 2 stained slides. (First & Last Slides are Stained)
Reactivity of Histo control slides are guaranteed for one year from the date of receipt and should be revalidated after one year to verify continued reactivity.
Please note the following: In human tissue unless otherwise indicated. Histopathology control slides are not quality controlled for IHC methods, as such, we cannot guarantee antigenicity.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR