(S)-(4,4',5,5',6,6'-Hexamethoxybiphenyl-2,2'-diyl)bis(diphenylphosphine) ≥97%
Supplier: Thermo Scientific Chemicals
047270-MD
047270-03
AA47270-03EA
399.99
USD
AA47270-03
AA47270-MD
(S)-(4,4',5,5',6,6'-Hexamethoxybiphenyl-2,2'-diyl)bis(diphenylphosphine) ≥97%
(S)-(4,4',5,5',6,6'-Hexamethoxybiphenyl-2,2'-diyl)bis(diphenylphosphine)
Formula:
C₄₂H₄₀O₆P₂ Storage Temperature: Ambient |
CAS Number:
133545-30-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...