SP Wilmad-LabGlass Multiple Distilling Receiver with 50mL Receiving Flasks, SP Industries
LG-6550-112 LG-6550-142 LG-6550-100 LG-6550-110 LG-6550-140 LG-6550-102
14204-732EA 128.09 USD
14204-732 14206-768 14207-748 14203-998 14206-644 14203-252
SP Wilmad-LabGlass Multiple Distilling Receiver with 50mL Receiving Flasks, SP Industries
These distilling heads consist of a vacuum adapter with inlet angled at 105° to the delivery stem, the distributing adapter, and four - 50mL receiving flasks with Standard Taper 24/40 outer joints.
  • Borosilicate glass
All pieces are connected using plastic clamps (LG-1004 included).