Low Constant Temperature Water Baths, Yamato
Supplier: Yamato
BBL-101-115V BBL-301-115V
76550-160EA 7422.75 USD
76550-160 76550-156
Low Constant Temperature Water Baths, Yamato
Baths Water Baths, Unstirred
This benchtop water bath features a wide operating range.
  • Pump circulation cooling/external circulation system
  • Temperature control range: −10~+80 °C (at room temperature 20 °C)
  • Temperature adjustment accuracy: ±0.1 °C
  • Operation functions: Fix temperature, auto stop, and auto start
  • Indicator for pump and refrigerator
Its larger tank space allows for more applications
A lid, drain hose, drain port, overflow hose, condenser filter, and bottom shelf are provided. The bath is also equipped with auto stop and start, RS485 communication function, temperature output terminal (4 to 20 mA), calibration offset, refrigerator pressure switch indicator, and advanced safety features such as self-diagnostic functions and an over-current circuit breaker.
Internal/external circulation is easy to change, and the shelf height is two-stage adjustable for different flask sizes. The refrigerator is a 160 W air-cooled, fully closed compressor.