Precision Constant Temperature Agitating Water Bath, Yamato
Supplier: Yamato
BK710 BK400 BA710 BK500 BA610 BK610 BA300 BK300 BA400 BA500
75875-982EA 8006.76 USD
75875-982 75875-976 75875-992 75875-978 75875-990 75875-980 75875-984 75875-974 75875-986 75875-988
Precision Constant Temperature Agitating Water Bath, Yamato
Baths Shaking Water Baths
This water bath has a high degree of temperature adjustment precision due to its high-precision thermostat and circulation pump.
  • Stirring Method: Stir by Pump
  • Operating Temp. Range: Room Temp +5~80°C
  • Digital Setting and Display
  • Observation Window
  • Fixed Temperature or Program Operation Available
The water bath has an interior of stainless steel and glass, and it has a temperature accuracy of ±0.3 °C.
A digital display indicates the temperature, time, and settings. Timer functions include auto start and stop and quick auto stop. Self-diagnostic features, an electric leakage breaker, and an overheat protector ensure safe operation. The unit includes plates, clamps, clamp holders, one tube connector, and a drainage cap.