Anti-IgM Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: MP Biomedicals
0855503
75877-366EA 612.94 USD
75877-366
Anti-IgM Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Fluorescein-Conjugated Goat IgG Fraction To Human IgM is the lyophilized powder of fluorescein-5-isothiocyanate (FITC "Isomer I")-conjugated goat IgG fraction to human IgM (5Fc µ) and buffer salts.

Immunoglobin M (IgM) antibody is prominent in early immune responses to most antigens and is the predominant response in certain antibody responses such as "natural" blood group antibodies. IgM is the major immunoglobin expressed on surface B lymphocytes. Most of the chronic lymphocytic leukemias, non Hodgkin lymphomas and Burkitt's lymphoma bear surface IgM, whereas plasmocytes from Waldenström's disease bear intracytoplasmic IgM.

Fluorescein-Conjugated Goat IgG Fraction To Human IgM is used as a reagent in immunofluorescence assays (IFA), cell staining (fluorescent microscopy and cell sorting), tissue staining, and blot immunostaining. (Note: F(ab')2 fragments are recommended for staining of cells or tissues which contain Fc receptors. Affinity purified antibodies are recommended to avoid non-specific binding from the inherent antibodies of host animals).

Antibody and FITC are conjugated under defined conditions to obtain the specified molar ratio (3-6 mol/mol). Conjugated protein is purified by gel filtration. The product is dialyzed into 0.02M sodium phosphate, 0.14M sodium chloride, pH 7.3, with 0.05% sodium azide, adjusted to standard titer, filtered through a 0.22 µm filter, vialed, and lyophilized.

The total protein is measured using the Biuret procedure with bovine albumin as the standard; F/P ratio is calculated using absorbance at 491.5nm. Antibody titer of the IgG fraction is standardized by equivalence-point precipitation. The IgG fraction is tested for purity and specificity at 40 mg/mL using immunoelectrophoresis. The product is mostly goat IgG; no trace of albumin is detected.

Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR