Sodium hypochlorite (8 - 10% Cl₂)

Danger

47285-A9
AA47285-A9EA 96.81 USD
AA47285-A9
Sodium hypochlorite (8 - 10% Cl₂)
Sodium hypochlorite
Formula: ClNaO
MW: 74.44 g/mol
Density: 1.2
Storage Temperature: Ambient
MDL Number: MFCD00011120
CAS Number: 7681-52-9
EINECS: 231-668-3
UN: 1791
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR