Ceftizoxime sodium ≥95%, Powder

Warning

J66554-MF J66554-MC
AAJ66554-MCEA 336.17 USD
AAJ66554-MC AAJ66554-MF
Ceftizoxime sodium ≥95%, Powder
Ceftizoxime sodium

A broad spectrum antibiotic targeting a wide variety of Gram-positive and Gram-negative bacteria

Formula: C₁₃H₁₂N₅NaO₅S₂
Melting Pt: 227 °C
Storage Temperature: Ambient
MDL Number: MFCD01682036
CAS Number: 68401-82-1

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR