AMG-9810

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (E)-3-(4-(tert-Butyl)phenyl)-N-(2,3-dihydrobenzo[b][1,4]dioxin-6-yl)acrylamide

J64202-MB
AAJ64202-MBEA 322.62 USD
AAJ64202-MB
AMG-9810
AMG-9810
Formula: C₂₁H₂₃NO₃
MW: 337.41 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00955103
CAS Number: 545395-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR