1-Boc-4-piperidone ≥97%

Warning

Synonyms: 4-Oxo-piperidine-1-carboxylic acidtert-butyl esterN-tert-BOC-4-PiperidinoneN-BOC-4-Piperidone

H37525-06 H37525-22 H37525-14
AAH37525-22EA 158 USD
AAH37525-22 AAH37525-06 AAH37525-14
1-Boc-4-piperidone ≥97%
1-Boc-4-piperidone
Formula: C₁₀H₁₇NO₃
MW: 199.25 g/mol
Melting Pt: 72…76°C
Storage Temperature: Ambient
MDL Number: MFCD00151800
CAS Number: 79099-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR