Campy FDA Agar
Supplier: Hardy Diagnostics
A139
75878-356PK 99.16 USD
75878-356
Campy FDA Agar
Media for Microbiology Prepared Media for Microbiology
CAMPY FDA is a modified Skirrows formula, similar to Blaser formula but with lysed horse blood instead of whole sheep blood, and PVANT antibiotics.
  • Modified Skirrows formula
  • Lysed horse blood