Fructose Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
EFRU-100
75878-340EA 543.21 USD
75878-340
Fructose Assay Kit, BioAssay Systems
Assays Cellular Assays
For quantitative determination of D-lactate (D-lactic acid) and evaluation of drug effects on its metabolism
  • Use as little as 20 μL samples. Linear detection range in 96-well plate: 12 to 1000 μM fructose.
  • Method: OD565nm
  • Samples: Serum, plasma, urine, saliva, milk, culture medium, food, beverage, agriculture etc
Fructose (C6H12O6, also called levulose or laevulose), is a monosaccharide found in honey, tree fruits, berries, melons, and some root vegetables along with glucose and galactose. The human body can use fructose for energy, however, too much consumption may lead to high triglycerides. Simple, direct and high-throughput assays for fructose determination find wide applications. BioAssay Systems’ reagent systems reacts directly and specifically with fructose to form a colored product. Glucose and galactose do not interfere. The color intensity at 565nm is directly proportional to the fructose concentration in the sample.