Acetonitrile, HiPerSolv CHROMANORM® suitable for LC-MS, UPLC, UHPLC, Ultra HPLC

Synonyms: Methane, cyano-Methyl cyanideEthanoic acid nitrileMethanecarbonitrileEthanenitrileCyanomethaneACN

BDHVBDH83642.201 BDHVBDH83642.050 BDHVBDH83642.202
BDH83642.201EA 10920.24 USD
BDH83642.201 BDH83642.202 BDH83642.050
Acetonitrile, HiPerSolv CHROMANORM® suitable for LC-MS, UPLC, UHPLC, Ultra HPLC
Acetonitrile

From the exclusive family of high-quality, competitively priced BDH Chemicals – High Purity solvents for HPLC applications.


Solvents are pumped through 0.2 micron filters on filling and packaged under nitrogen.


Ordering information: To purchase these solvents in stainless steel returnable drums as part of the VWR ReCAP program, please contact your local VWR sales representative. A separate contract for VWR ReCAP products is required.

Formula: H₃CCN
MW: 41.05 g/mol
MDL Number: MFCD00001878
CAS Number: 75-05-8
UN: 1648
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/89/10/718910.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR