Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Plasminogen, Human Plasma, MilliporeSigma

Supplier: MilliporeSigma
Calbiochem®
528175-120U
Plasminogen is a native plasminogen from human plasma. Plasminogen is a single chain glycoprotein found in normal plasma at ~12 mg per deciliter and is inactive precursor of serine protease plasmin.

  • Lyophilized form
  • Synonyms: Profibrinolysin, Plasma Trypsinogen
  • Purity: ≥95%
  • Specific Activity: ≥120 units/mg protein

Prepared from plasma that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.




 

ORDER

Expand All / Collapse All
Packaging Supplier No. VWR Catalog Number Unit Price Quantity
Glass bottle 528175-120U 80053-738 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/Peptide Name Plasminogen
Protein/Peptide Type Native
Species Human
Source Plasma
Purity ≥95%
Formulation Lyophilized from 20 mM Tris-HCl, pH 7.5, with 2 mM EDTA.
Storage Conditions -20°C
Shipping Temperature Ambient
Protein Type Native
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty