1-Hexadecanol (cetyl alcohol) ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Palmityl alcoholCetyl alcohol

4016001GM
100291-076EA 189.62 USD
100291-076
1-Hexadecanol (cetyl alcohol) ≥99%
1-Hexadecanol

Storage conditions: Store in a cool, dry place

Formula: H₃C(CH₂)₁₄CH₂OH
MW: 242.45 g/mol
MDL Number: MFCD00004760
CAS Number: 36653-82-4
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/71/21/21287121.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR