Benzyl-1-(oxetan-3-yl)piperidin-4-ylcarbamate ≥95%

Supplier: Matrix Scientific

115833-500MG 115833-5G
102900-292EA 1318.46 USD
102900-292 102900-296
Benzyl-1-(oxetan-3-yl)piperidin-4-ylcarbamate ≥95%
Benzyl-1-(oxetan-3-yl)piperidin-4-ylcarbamate

Hazard: Irritant

Formula: C₁₆H₂₂N₂O₃
MW: 290.37 g/mol
MDL Number: MFCD20233246
CAS Number: 1349716-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR