5,5-Bis(hydroxymethyl)-2-phenyl-1,3-dioxane ≥95%

Supplier: Matrix Scientific

Synonyms: MonobenzalpentaerythritolO,O'-Benzylidenepentaerythritol(2-Phenyl-1,3-dioxane-5,5-diyl)dimethanol

115975-5G
102900-018EA 413.85 USD
102900-018
5,5-Bis(hydroxymethyl)-2-phenyl-1,3-dioxane ≥95%
5,5-Bis(hydroxymethyl)-2-phenyl-1,3-dioxane

Hazard: Irritant

Formula: C₁₂H₁₆O₄
MW: 224.26 g/mol
MDL Number: MFCD00085385
CAS Number: 2425-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR