Ethyl-5-(trifluoromethyl)-1-[3-(trifluoromethyl)phenyl]-1H-pyrazole-4-carboxylate ≥97%

Supplier: Matrix Scientific

114687-1G
102894-528EA 738.46 USD
102894-528
Ethyl-5-(trifluoromethyl)-1-[3-(trifluoromethyl)phenyl]-1H-pyrazole-4-carboxylate ≥97%
Ethyl-5-(trifluoromethyl)-1-[3-(trifluoromethyl)phenyl]-1H-pyrazole-4-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₀F₆N₂O₂
MDL Number: MFCD28099758
CAS Number: 112055-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR