Methyl-5-methyl-2-phenyl-1,3-oxazole-4-carboxylate ≥97%

Supplier: Matrix Scientific

114686-1G
102894-526EA 813.85 USD
102894-526
Methyl-5-methyl-2-phenyl-1,3-oxazole-4-carboxylate ≥97%
Methyl-5-methyl-2-phenyl-1,3-oxazole-4-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₁NO₃
MDL Number: MFCD08870502
CAS Number: 100063-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR