(2R)-1-(tert-Butoxycarbonyl)-4-hydroxypyrrolidine-2-carboxylic acid ≥97%

Supplier: Matrix Scientific

114548-1G
102894-272EA 1443.08 USD
102894-272
(2R)-1-(tert-Butoxycarbonyl)-4-hydroxypyrrolidine-2-carboxylic acid ≥97%
(2R)-1-(tert-Butoxycarbonyl)-4-hydroxypyrrolidine-2-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₁₇NO₅
MDL Number: MFCD09756586
CAS Number: 1157846-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR