VeriSpec® Instrument Check Standard, 5 Components, RICCA Chemical Company
Supplier: RICCA Chemical Company
VeriSpec®
RV010783-100N
RCRV010783100NEA 245.35 USD
RCRV010783100N
VeriSpec® Instrument Check Standard, 5 Components, RICCA Chemical Company
Standards Cationic Standards Multi-Element Standards
Certified reference material (CRM) for ICP-MS
10 ppm: Mo, Th, U
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility