VeriSpec® Thorium Standard for ICP/MS 10 ppm in 2% HNO₃
Supplier: Ricca Chemical
VeriSpec™
RV010628-50N
RCRV010628-50NEA 61.59 USD InStock
RCRV010628-50N
VeriSpec® Thorium Standard for ICP/MS 10 ppm in 2% HNO₃
Standards Cationic Standards Thorium Standards
Certified reference material (CRM) for ICP-MS
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility