VeriSpec® Rhenium Standard for ICP 10 g/L in H₂O, RICCA Chemical Company
Supplier: RICCA Chemical Company
VeriSpec™
RV010408-50N RV010408-100N RV010408-500N RV010408-250N
RCRV010408100NEA 1357.48 USD
RCRV010408100N RCRV010408250N RCRV010408-50N RCRV010408500N
VeriSpec® Rhenium Standard for ICP 10 g/L in H₂O, RICCA Chemical Company
Standards Cationic Standards Rhenium Standards
Certified reference material (CRM) for ICP
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Caution: 10 g/L= 10,000 µg/mL