VeriSpec™ Potassium Standard for ICP 10 g/L in H₂O, RICCA Chemical Company
Supplier: RICCA Chemical Company
VeriSpec™
RV010390-100N RV010390-500N RV010390-50N RV010390-250N
RCRV010390-50NEA 89.57 USD
RCRV010390-50N RCRV010390250N RCRV010390100N RCRV010390500N
VeriSpec™ Potassium Standard for ICP 10 g/L in H₂O, RICCA Chemical Company
Standards Cationic Standards Potassium Standards
Certified reference material (CRM) for ICP
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Caution: 10 g/L= 10,000 µg/mL