VeriSpec™ Potassium Ionization Buffer for AAS 2% K in 1% HCl, RICCA Chemical Company
Supplier: RICCA Chemical Company
VeriSpec™
RV010143-100N
RCRV010143100NEA 329.39 USD
RCRV010143100N
VeriSpec™ Potassium Ionization Buffer for AAS 2% K in 1% HCl, RICCA Chemical Company
Standards Cationic Standards Potassium Standards
Certified reference material (CRM) for AA (atomic absorption)
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility