VeriSpec® Calcium (Ca) Standard for AAS 1000 ppm in 12% Ethanol, RICCA Chemical Company
Supplier: RICCA Chemical Company
VeriSpec™
RV010116-100N RV010116-500N
RCRV010116500NEA 223.86 USD
RCRV010116500N RCRV010116100N
VeriSpec® Calcium (Ca) Standard for AAS 1000 ppm in 12% Ethanol, RICCA Chemical Company
Standards Cationic Standards Calcium Standards
Certified reference material (CRM) for AA (atomic absorption)
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility