VeriSpec™ Potassium Ionization Buffer for AAS 2% K in 1% HNO₃, RICCA Chemical Company
Supplier: RICCA Chemical Company
VeriSpec™
RV010010-500N RV010010-100N
RCRV010010500NEA 975.89 USD
RCRV010010500N RCRV010010100N
VeriSpec™ Potassium Ionization Buffer for AAS 2% K in 1% HNO₃, RICCA Chemical Company
Standards Cationic Standards Potassium Standards
Certified reference material (CRM) for AA (atomic absorption)
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility