SP Wilmad-LabGlass Hempel Distilling Columns, SP Industries
Supplier: SP Industries
LG-5791-106 LG-5791-126 LG-5791-120
14205-252EA 600.08 USD
14205-252 14206-194 14204-980
SP Wilmad-LabGlass Hempel Distilling Columns, SP Industries
These Hempel Columns consist of a plain tube with full length inner joint at bottom and outer joint of the same size at top.
  • Made of borosilicate glass
Column section is enclosed in evacuated jacket. Near bottom there is a row of indentations to support random type packing material.