Would you like to log in again?
Straight Sided Round Jars, Borosilicate Glass, Clear, Qorpak®
Supplier: QORPAK
273695 271012 268240
Clear Straight Sided Round Jars are now available in Borosilicate Glass, designed for use in critical applications where thermal stability, chemical corrosion resistance, and durability are required.
These jars are designed without shoulders for maximum storage capacity and content visibility. Borosilicate glass jars are more resistant to temperature change than regular glass, and provide greater assurance of content integrity. Borosilicate glass jars can be quickly identified by the Qorpak® logo on the bottom of the jar.
ORDER
Expand All / Collapse All
Description Capacity Cap Size Cap Type Supplier No. VWR Catalog Number Unit Price Quantity
Clear Type I Borosilicate Glass Straight Sided Jar, Cap Attached 237 mL (8 oz.) 70-400 White Polypropylene F217 Liner 273695 75871-670 Case of 12 Retrieving
Clear Type I Borosilicate Glass Straight Sided Jar, Cap Attached 237 mL (8 oz.) 70-400 Green Thermoset F217 & Fluoropolymer Resin Liner 271012 75871-668 Case of 12 Retrieving
Clear Type I Borosilicate Glass Straight Sided Jar 237 mL (8 oz.) 70-400 Bottle Only 268240 75871-638 Case of 12 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty