UVP GelStudio PLUS Gel Documentation Imaging Systems, Analytik Jena
Supplier: Analytik Jena
849-00552-4 849-00552-2 849-00553-4 849-00553-4 849-00552-4 849-00553-2
75873-984EA 13332.3 USD
75873-984 CA75928-110 75873-980 CA75927-894 CA75928-108 CA75927-896
UVP GelStudio PLUS Gel Documentation Imaging Systems, Analytik Jena
Photodocumentation Systems
The UVP GelStudio imaging systems extensive analytical software and range of epi LEDs included with the system ensure every application need is met.
  • Ideal for imaging and documentation of DNA and Protein gels, blue/white excitable dyes, TLC plates, colorimetry, colony plates, and a wide range of fluorescent dyes
  • Acquire high quality images with an efficient photon-to-signal conversion 5 MP camera
  • Wide aperture lens optics, 8-48mm f/1.2, capture more light in low-light applications. Signals come faster and are stronger. This guarantees extremely fast image capture
  • Available as either a PC-operated unit or as a stand-alone instrument with an integrated color touchscreen
  • Overhead white, green, red and blue LEDs come as standard in the series
  • Includes unlimited copies of VisionWorks Software, with comprehensive features, optimizes image acquisition and analysis
  • Unmodified raw data is collected, giving users the choice of applying image enhancement tools
The GelStudio Series by Analytik Jena is the next generation of digital imagers, a direct response to the expanding needs of researchers. The UVP GelStudio product family features the newest technology, a uniquely intuitive software interface and a top-of-the-line 5MP gel imaging camera, giving you uncompromised data, every single time.
Multiple illumination sources. All GelStudio systems come with a 302 nm UV transilluminator and overhead RGBW LEDs as standard enabling a wide scope of fluorescent and nonfluorescent imaging applications. The included Ethidium Bromide emission filter allows for imaging of the most common applications; optional accessories such as additional emission filters, conversion screens and epi-UV sources add further versatility to the systems on demand.
High performance cameras and software. Optimize your application with the use of the UVP GelStudio’s extremely sensitive camera to deliver uncompromised raw data, promising the highest quantitative value. All imagers are operated using VisionWorks Software, guaranteeing powerful capture, quantitating and analysis. Application-based icons for automation, which are included in the software package, offer one-touch capture. The software allows for creating custom icons and workflows based on users’ needs. Extensive image enhancement and analysis tools give users control to personalize their experiments and make use of enhancement and annotation features including noise reduction, background subtraction, inversion, pseudocolor, compositing and more.
Certifications: CSA certified. CE compliant.
Ordering information: IQ/OQ (Installation and Qualification) Documents and on-site support can be purchased. Three-year warranty. All systems include multiple copies of the full package VisionWorks Acquisition and Analysis Software, empty flash drive, gel tray, gel cutter and an ethidium bromide filter. An integrated tablet with 13.3" multi-touch color screen, keyboard, mouse, and stylus are included with the Touch version only.
Additional filters are available for purchase. Accessories not included. Additional lighting options: UVP LED Visi-White Light plate, UVP Visi-Blue Plate. An optional compact Thermal Printer is also available to generate archive quality, 256 gray scale prints. Contact your VWR representative for emission and excitation filter sets.
IQ/OQ (Installation and Qualification) Documents and on-site support can be purchased.