VWR® PCR Plates for Roche® Lightcycler®
Supplier: VWR International
3455-40-521AV
10755-256CS 697.1 USD
10755-256
VWR® PCR Plates for Roche® Lightcycler®
Microplates PCR Plates
PCR plates made of polypropylene for Roche® Lightcycler® are certified free of Endotoxin, Rnase/Dnase, and ATP.