You Searched For: 4-(2-Dimethylaminoethoxy)benzaldehyde
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Supplier: Ambeed
Description: 4-(2-(Dimethylamino)ethoxy)benzaldehyde, Purity: 97%, CAS Number: 15182-92-0, Appearance: Light-yellow to yellow to yellow-brown liquid, Storage: Inert atmosphere, 2-8C, Size: 5G