You Searched For: Ethyl-2-(3-oxo-2-piperazinyl)acetate
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Supplier: Ambeed
Description: Ethyl 2-(3-oxopiperazin-2-yl)acetate, Purity: 98%, CAS Number: 33422-35-4, Appearance: White to yellow powder or crystals, Storage: Keep in dark place, Sealed in dry, 2-8 C, Size: 250mg