You Searched For: 1,2,4,5-Tetraiodobenzene
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Supplier: Ambeed
Description: 1,2,4,5-Tetraiodobenzene, Purity: 95%, CAS Number: 636-31-7, Appearance: Yellow to red to brown Powder or crystals, Storage: Keep in dark place, Sealed in dry, Room Temperature, Size: 1g