You Searched For: Iodine (iodine-iodide)


13  results were found

SearchResultCount:"13"

Sort Results

List View Easy View

Rate These Search Results

Supplier: Ricca Chemical
Description: 1 mL = 0.4008 mg S2-
Catalog Number: (RC39901)
Supplier: Ricca Chemical
Description: ASTM D1510 for iodine absorption number of carbon black. Contains Max. 1% iodine and Max. 6% potassium iodide.

Supplier: Ricca Chemical
Description: IODINE 0.020 NORMAL (N/50)
Supplier: Ricca Chemical
Description: 1 mL = 1 mg Cl2
Supplier: Ricca Chemical
Description: 0.100 (+/-0.002) Normal. Meets EPA Specifications. Container: Glass.
Catalog Number: (RC400232)
Supplier: Ricca Chemical
Description: IODINE 0.1563 NORMAL 32OZ

SDS Small Business Enterprise


Supplier: Ricca Chemical
Description: IODINE 0.0100 NORMAL (N/100)
Catalog Number: (RC3992-1)
Supplier: Ricca Chemical
Description: IODINE 0.0624 NORMAL 1GA EA

Supplier: Ricca Chemical
Description: Contains 1.2–13.0% iodine and 4–25% potassium iodide. Standardized at 25°C against chemicals whose certification is traceable to NIST.
Supplier: Ricca Chemical
Description: IODINE 0.200 N BOTTLE 500ML
Catalog Number: (RC3985-1)
Supplier: Ricca Chemical
Description: (Iodine-Iodide) 0.0315n

Catalog Number: (SX183134)
Supplier: SPEX CERTIPREP LLC
Description: Iodine solution is a Carbon Black reagent designed for ASTM D1510.


Supplier: Electron Microscopy Sciences
Description: Weigert's iodine solution is a histological stain. Demonstration of connective tissue is useful in cases of emphysema, arteriosclerosis and other vasc...

Minority or Woman-Owned Business Enterprise

Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 13 of 13
no targeter for Bottom