Would you like to log in again?
  • Criteria
  • Supplier
  • Suppliers found in search results
    Sort by:
  • Search Within Results
You Searched for: "petroleumbenzin-20-40"
SearchPresentationType-VERTICAL
SearchResultCount:"1"
Sort by:
Supplier:  Avantor Performance Materials
Description:   Ligroin; ether, petroleum; naphtha. CAS RN 8032-32-4. Density: 0.62 kg/L. ‘BAKER ANALYZED’* Reagent. Boiling Range: 20-40[degree]C. Lot analysis on label. Packaged in a SAFETAINER*. 4L.